Merck
LC/MS Analysis of L-Methionine Sulfoximine, Sulfone and Sulfoxide on Ascentis® Express HILIC
LC/MS Analysis of L-Methionine Sulfoximine, Sulfone and Sulfoxide on Ascentis® Express HILIC application for HPLC
材料
分析柱
产品编号
说明
价格
Ascentis® Express HILIC, 2.7 μm HPLC色谱柱
2.7 μm particle size, L × I.D. 10 cm × 3 mm
標準品
产品编号
说明
价格
L -蛋氨酸砜
L -甲硫氨酸亚砜
L-蛋氨酸亚砜酰亚胺
suitable for cell culture
CONDITIONS
column
Ascentis Express HILIC, 10 cm x 3.0 mm I.D., 2.7 μm particles (53970-U)
mobile phase
[A] acetonitrile; [B] water with 0.1% formic acid, pH 3.5 with ammonium formate; (75:25, A:B)
flow rate
0.4 mL/min
pressure
1350 psi 93 bar
temp.
25 °C
detector
MS, ESI (+), TIC, m/z 150-300
injection
5 μL
sample
10 μg/mL in 90:10, methanol:water
说明
分析報告
L-methionine sulfoximine and L-buthionine sulfoximine are used in are used to prevent additional enzyme activity in Chinese hampster ovary (CHO) cell lines with supplemental Glutamine Synthase (GS). It is of interest to be able to monitor both purity of such molecules in order to control addition to cell media as well as for the assay of parent molecules during use. The analytes are highly polar and thus should be amenable to HILIC-LC-MS analysis. This first installment investigates the ability of Ascentis Express HILIC and Ascentis Express OH5 stationary phases for the separation of the methionine analog and related compounds as well as methionine and buthionine separations.