Merck
HPLC Analysis of 3-Phenyllactic Acid Enantiomers on Astec® CHIROBIOTIC® R
HPLC Analysis of 3-Phenyllactic Acid Enantiomers on Astec® CHIROBIOTIC® R application for HPLC
材料
分析色谱柱
产品编号
说明
价格
Astec® CHIROBIOTIC® R手性HPLC柱
5 μm particle size, L × I.D. 25 cm × 4.6 mm
标准
产品编号
说明
价格
3-苯基乳酸
≥98%
D -(+)-3-苯乳酸
98%
L-(-)-3-苯基乳酸
98%
CONDITIONS
column
CHIROBIOTIC R, 25 cm x 4.6 mm I.D., 5 μm particles (13024AST)
mobile phase
0.1% ammonium acetate in methanol
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 230 nm
injection
0.5 μL