Merck
GC Analysis of 1,2-Alkyldiol Enantiomers (O-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX G-TA
GC Analysis of 1,2-Alkyldiol Enantiomers (O-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX™ G-TA suitable for GC
材料
分析色谱柱
产品编号
说明
价格
Astec® CHIRALDEX G-TA 毛细管GC色谱柱
L × I.D. 30 m × 0.25 mm, df 0.12 μm
标准
产品编号
说明
价格
1,2-丙二醇
puriss. p.a., ACS reagent, ≥99.5% (GC)
1,2-丙二醇
tested according to Ph. Eur.
1,2-丙二醇
meets analytical specification of Ph. Eur., BP, USP, ≥99.5%
1,2-丙二醇
analytical standard
(R)-(-)-1,2-丙二醇
96%
(S)-(+)-1,2-丙二醇
96%
1,2-丙二醇-d2
98 atom % D
DL-1,2-己二醇
98%
1,2-辛二醇
98%
1,2-丙二醇-d8
98 atom % D
1,2-丙二醇
ACS reagent, ≥99.5%
1,2-戊二醇
96%
1,2-丙二醇
ReagentPlus®, 99%
1,2-丙二醇-d6
98 atom % D
1,2-丙二醇-1,2-13C2
99 atom % 13C
1,2-丙二醇
SAJ first grade, ≥99.0%
1,2-丙二醇
SAJ special grade, ≥99.0%
(R)-(+)-1,2-Octanediol
≥95.0% (sum of enantiomers, GC)
CONDITIONS
column
Astec CHIRALDEX G-TA, 30 m x 0.25 mm I.D., 0.12 μm (73033AST)
oven
80 °C, 5 °C/min to 130 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 30 psi
injection
1 μL, 80:1 split
sample
various alkyldiol enantiomers (O-TFA derivatives)
说明
法律信息
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC