跳转至内容
Merck
  • Alpha 2-adrenoceptors and I1-imidazoline binding sites: relationship with catecholamines in women of reproductive age.
Alpha 2-adrenoceptors and I1-imidazoline binding sites: relationship with catecholamines in women of reproductive age.
Journal of psychiatric research (1998-08-07)
J E Piletz, M Andrew, H Zhu, Y Z Feng, J Rains, A Halaris
摘要
A comparison is presented between plasma catecholamine concentrations and platelet [125I]-p-iodoclonidine binding sites in 16 healthy women. Blood samples were obtained at six regularly spaced intervals over two consecutive menstrual cycles from healthy women with regular menstrual periods. Although no cycle-related changes were observed per se, there were significant correlations between the platelet binding sites and plasma norepinephrine and epinephrine concentrations. The densities of platelet alpha 2-adrenoceptors were negatively correlated in an exponential fashion (r2 = 0.694, P = 0.009) with plasma epinephrine concentrations, implying agonist-induced downregulation. On the other hand, platelet I1-imidazoline binding sites were positively correlated with plasma concentrations of norepinephrine in a linear fashion (r2 = 0.326, P = 0.021). This is the first indication that I1 binding sites might be upregulated by a physiological factor. Furthermore, the data suggest that elevations in plasma norepinephrine might explain reports of upregulated I1 binding sites in depressed patients.
材料
货号
品牌
产品描述
Supelco
对碘可乐定 盐酸盐, analytical standard, for drug analysis