跳转至内容
Merck
  • Combination of microwave assisted micellar extraction and liquid chromatography for the determination of organophosphorous pesticides in soil samples.
Combination of microwave assisted micellar extraction and liquid chromatography for the determination of organophosphorous pesticides in soil samples.
Journal of chromatography. A (2005-07-13)
C Padrín-Sanz, R Halko, Z Sosa-Ferreraa, J J Santana-Rodríguez
摘要
A new methodology based on the microwave assisted micellar extraction (MAME) technique has been optimised, using soil samples, to extract and determine a mixture of the eight organophosphorous pesticides mainly used in agriculture. The pesticides under study have been extracted using the non-ionic surfactants polyoxyethylene 10 lauryl ether (POLE) and oligoethylene glycol monoalkyl ether (Genapol X-080). The optimal extraction variables, such as surfactant concentration, pH, radiation time and microwave power were determined for each surfactant. The results show the advantage of using POLE instead of Genapol X-080 for the extraction of the organophosphorus pesticides with recoveries higher than 70% for most of the compounds and relative standard deviations (RSD) below 2.6%. This method was successfully applied to fresh samples as well as to aged samples for the analysis of soils with different characteristics and compared with the traditional Soxhlet technique.
材料
货号
品牌
产品描述
Sigma-Aldrich
Genapol® X-080
Sigma-Aldrich
Genapol ® X-100