跳转至内容
Merck
  • Selective pressurised liquid extraction of halogenated pesticides and polychlorinated biphenyls from pine needles.
Selective pressurised liquid extraction of halogenated pesticides and polychlorinated biphenyls from pine needles.
Journal of chromatography. A (2012-09-04)
Karen S Lavin, Kimberly J Hageman
摘要
Pine needles are an ideal matrix for the long-term monitoring of semi-volatile organic compounds in the atmosphere because they are naturally occurring and distributed worldwide. However, typical extraction methods result in the co-elution of matrix-interfering compounds and thus require time-consuming clean-up steps. A selective pressurised liquid extraction method (S-PLE) for extracting current-use pesticides, historic-use pesticides and polychlorinated biphenyls (PCBs) from pine needles that does not require additional clean-up steps was developed and validated. The selective extraction was achieved by packing the extraction vessel with a fat retainer, Florisil, down-stream of the pine needle matrix. A fat to fat-retainer ratio of 0.0057 (corresponding to 35 g of Florisil per 10 g of pine needle sample) was selected to minimise the co-elution of matrix-interfering compounds. Three 5-min extractions were performed using 25:75 (v/v) dichloromethane:n-hexane and a solvent flush of 150%. The mean recovery of spiked current-use pesticides, historic-use pesticides and PCBs using this method was 71%, 72%, and 84%, respectively. The method was validated by comparing target analyte concentrations measured in a pine needle sample using the optimised S-PLE method to those obtained using a conventional PLE method with external clean-up. The method detection limits and reproducibility were similar for the two methods; however, sample preparation time was 67% shorter when using the S-PLE method.
材料
货号
品牌
产品描述
Sigma-Aldrich
正己烷, suitable for HPLC, ≥97.0% (GC)
Sigma-Aldrich
正己烷, suitable for HPLC, ≥95%
Sigma-Aldrich
正己烷, ReagentPlus®, ≥99%
Sigma-Aldrich
正己烷, Laboratory Reagent, ≥95%
Sigma-Aldrich
正己烷, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
正己烷, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
正己烷, JIS special grade, ≥96.0%
Sigma-Aldrich
正己烷, anhydrous, 95%
Supelco
正己烷, analytical standard
Sigma-Aldrich
正己烷, SAJ first grade, ≥95.0%
Sigma-Aldrich
正己烷, suitable for HPLC
Sigma-Aldrich
正己烷, JIS 300, ≥96.0%, for residue analysis
Sigma-Aldrich
正己烷, for residue analysis, JIS 5000
Sigma-Aldrich
正己烷, JIS 1000, ≥96.0%, for residue analysis
Sigma-Aldrich
正己烷, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
正己烷, ≥96.0%, suitable for residual phthalate analysis