跳转至内容
Merck
  • Constituents and secondary metabolite natural products in fresh and deteriorated cassava roots.
Constituents and secondary metabolite natural products in fresh and deteriorated cassava roots.
Phytochemistry (2010-02-09)
Soad A L Bayoumi, Michael G Rowan, John R Beeching, Ian S Blagbrough
摘要
A phytochemical analysis of cassava (Manihot esculenta Crantz) fresh roots and roots suffering from post-harvest physiological deterioration (PPD) has been carried out. The first isolation and identification of galactosyl diacylglycerides from fresh cassava roots is reported, as well as beta-carotene, linamarin, and beta-sitosterol glucopyranoside. The hydroxycoumarin scopoletin and its glucoside scopolin were identified from cassava roots during PPD, as well as trace quantities of esculetin and its glucoside esculin. There is no isoscopoletin in cassava roots during PPD.
材料
货号
品牌
产品描述
Sigma-Aldrich
七叶苷水合物 水合物, ≥98%
Sigma-Aldrich
七叶苷水合物 水合物, ≥98.0% (HPLC)
Sigma-Aldrich
α-羟基异丁腈 β- D -吡喃葡萄糖苷, ≥97% (HPLC)