跳转至内容
Merck
  • A fluorogenic near-infrared imaging agent for quantifying plasma and local tissue renin activity in vivo and ex vivo.
A fluorogenic near-infrared imaging agent for quantifying plasma and local tissue renin activity in vivo and ex vivo.
American journal of physiology. Renal physiology (2012-06-08)
Jun Zhang, Dorin V Preda, Kristine O Vasquez, Jeff Morin, Jeannine Delaney, Bagna Bao, M David Percival, Daigen Xu, Dan McKay, Michael Klimas, Bohumil Bednar, Cyrille Sur, David Z Gao, Karen Madden, Wael Yared, Milind Rajopadhye, Jeffrey D Peterson
摘要
The renin-angiotensin system (RAS) is well studied for its regulation of blood pressure and fluid homeostasis, as well as for increased activity associated with a variety of diseases and conditions, including cardiovascular disease, diabetes, and kidney disease. The enzyme renin cleaves angiotensinogen to form angiotensin I (ANG I), which is further cleaved by angiotensin-converting enzyme to produce ANG II. Although ANG II is the main effector molecule of the RAS, renin is the rate-limiting enzyme, thus playing a pivotal role in regulating RAS activity in hypertension and organ injury processes. Our objective was to develop a near-infrared fluorescent (NIRF) renin-imaging agent for noninvasive in vivo detection of renin activity as a measure of tissue RAS and in vitro plasma renin activity. We synthesized a renin-activatable agent, ReninSense 680 FAST (ReninSense), using a NIRF-quenched substrate derived from angiotensinogen that is cleaved specifically by purified mouse and rat renin enzymes to generate a fluorescent signal. This agent was assessed in vitro, in vivo, and ex vivo to detect and quantify increases in plasma and kidney renin activity in sodium-sensitive inbred C57BL/6 mice maintained on a low dietary sodium and diuretic regimen. Noninvasive in vivo fluorescence molecular tomographic imaging of the ReninSense signal in the kidney detected increased renin activity in the kidneys of hyperreninemic C57BL/6 mice. The agent also effectively detected renin activity in ex vivo kidneys, kidney tissue sections, and plasma samples. This approach could provide a new tool for assessing disorders linked to altered tissue and plasma renin activity and to monitor the efficacy of therapeutic treatments.
材料
货号
品牌
产品描述
Sigma-Aldrich
Cathepsin G from human leukocytes, lyophilized powder, ≥60 units/mg protein (Bradford)