跳转至内容
Merck
  • Proteomics reveals Rictor as a noncanonical TGF-β signaling target during aneurysm progression in Marfan mice.
Proteomics reveals Rictor as a noncanonical TGF-β signaling target during aneurysm progression in Marfan mice.
American journal of physiology. Heart and circulatory physiology (2018-07-14)
Sarah J Parker, Aleksandr Stotland, Elena MacFarlane, Nicole Wilson, Amanda Orosco, Vidya Venkatraman, Kyle Madrid, Roberta Gottlieb, Harry C Dietz, Jennifer E Van Eyk
摘要
The objective of the present study was to 1) analyze the ascending aortic proteome within a mouse model of Marfan syndrome (MFS; Fbn1C1041G/+) at early and late stages of aneurysm and 2) subsequently test a novel hypothesis formulated on the basis of this unbiased proteomic screen that links changes in integrin composition to transforming growth factor (TGF)-β-dependent activation of the rapamycin-independent component of mammalian target of rapamycin (Rictor) signaling pathway. Ingenuity Pathway Analysis of over 1,000 proteins quantified from the in vivo MFS mouse aorta by data-independent acquisition mass spectrometry revealed a predicted upstream regulator, Rictor, that was selectively activated in aged MFS mice. We validated this pattern of Rictor activation in vivo by Western blot analysis for phosphorylation on Thr1135 in a separate cohort of mice and showed in vitro that TGF-β activates Rictor in an integrin-linked kinase-dependent manner in cultured aortic vascular smooth muscle cells. Expression of β3-integrin was upregulated in the aged MFS aorta relative to young MFS mice and wild-type mice. We showed that β3-integrin expression and activation modulated TGF-β-induced Rictor phosphorylation in vitro, and this signaling effect was associated with an altered vascular smooth muscle cell proliferative-migratory and metabolic in vitro phenotype that parallels the in vivo aneurysm phenotype in MFS. These results reveal that Rictor is a novel, context-dependent, noncanonical TGF-β signaling effector with potential pathogenic implications in aortic aneurysm. NEW & NOTEWORTHY We present the most comprehensive quantitative analysis of the ascending aortic aneurysm proteome in Marfan syndrome to date resulting in novel and potentially wide-reaching findings that expression and signaling by β3-integrin constitute a modulator of transforming growth factor-β-induced rapamycin-independent component of mammalian target of rapamycin (Rictor) signaling and physiology in aortic vascular smooth muscle cells.
材料
货号
品牌
产品描述
Sigma-Aldrich
抗-纤连蛋白 兔抗, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
抗磷酸化-Smad2(Ser465/467)抗体,克隆A5S,兔单克隆, culture supernatant, clone A5S, Upstate®