跳转至内容
Merck
  • HES1 is a master regulator of glucocorticoid receptor-dependent gene expression.
HES1 is a master regulator of glucocorticoid receptor-dependent gene expression.
Science signaling (2013-12-05)
Javier R Revollo, Robert H Oakley, Nick Z Lu, Mahita Kadmiel, Maheer Gandhavadi, John A Cidlowski
摘要
Hairy and enhancer of split-1 (HES1) is a basic helix-loop-helix transcription factor that is a key regulator of development and organogenesis. However, little is known about the role of HES1 after birth. Glucocorticoids, primary stress hormones that are essential for life, regulate numerous homeostatic processes that permit vertebrates to cope with physiological challenges. The molecular actions of glucocorticoids are mediated by glucocorticoid receptor-dependent regulation of nearly 25% of the genome. Here, we established a genome-wide molecular link between HES1 and glucocorticoid receptors that controls the ability of cells and animals to respond to stress. Glucocorticoid signaling rapidly and robustly silenced HES1 expression. This glucocorticoid-dependent repression of HES1 was necessary for the glucocorticoid receptor to regulate many of its target genes. Mice with conditional knockout of HES1 in the liver exhibited an expanded glucocorticoid receptor signaling profile and aberrant metabolic phenotype. Our results indicate that HES1 acts as a master repressor, the silencing of which is required for proper glucocorticoid signaling.
材料
货号
品牌
产品描述
Sigma-Aldrich
EZ-Magna ChIP® A -染色质免疫沉淀试剂盒, Single day chromatin immunoprecipitation (ChIP) kit containing all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions using magnetic A beads. Control primers included.