返回
J-KEM
®
basic temperature controller
Products
品牌
Sigma-Aldrich (5)
筛选产品
Showing 1-5 of 5
比较
货号
说明
价格
Z284718
J-KEM
®
basic temperature controller,
Model 310, temperature -50-200 °C
展开
查看定价
Z210226
J-KEM
®
basic temperature controller,
Model 210, temperature -200-250 °C
展开
查看定价
Z248207
J-KEM
®
basic temperature controller,
Model 210, temperature -50-200 °C
展开
查看定价
Z248215
J-KEM
®
basic temperature controller,
Model 210/T, temperature -200-250 °C
展开
查看定价
Z248223
J-KEM
®
basic temperature controller,
Model 210/T, temperature -50-200 °C