返回
3D Biotek Polycaprolactone (PCL) scaffold inserts
Products
产品分类
inserts (3)
制造商
吸附类型
孔径(µm)
无菌性
筛选产品
Showing 1-3 of 3
比较
货号
说明
价格
Z694673
3D Biotek Polycaprolactone (PCL) scaffold inserts,
for 3D bioreactor 24 well chamber, fiber diam. ~300 μm
展开
查看定价
Z694657
3D Biotek Polycaprolactone (PCL) scaffold inserts,
fiber diam. ~300 μm, ~ 300 μm spacing
展开
查看定价
Z694665
3D Biotek Polycaprolactone (PCL) scaffold inserts,
for 3D bioreactor 12 well chamber, fiber diam. ~300 μm, ~ 300 μm spacing