2637525
1 of 10
Supelco
2637505
进样口衬管,直接 (SPME) 型,直型设计(未包装)
2637501
2633901
进样口衬管,固相微萃取/不分流,用于带 SPL-17 进样器的 17A
20654
Thermogreen® LB-2隔垫,固体圆盘
21003-U
Therm-O-Ring 密封圈
57354-U
Carbowax-聚乙二醇 (PEG) 固相微萃取萃取头
21004-U
57330-U
SPME 光纤支架
2633905
28336-U
模塑 Thermogreen ® LB-2 隔膜,带注射孔
pkg of 25 ea
78.5 mm × 6.5 mm × 0.75 mm
正在寻找类似产品? 访问 产品对比指南
默克Supelco®_气相色谱柱_配件和吸附剂-默克生命科学
Choose from one of the most recent versions:
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact 客户支持