2630901
别名:
进样口衬管,分流式(未填充),用于 AutoSystem
1 of 10
Supelco
33321-U
用于 Waters 96 孔转盘的 8×40mm 样品瓶
Sigma-Aldrich
Z515620
Aldrich® evaporating flask, pear-shaped
Millipore
1.46452
HAYFLICK液体培养基
CLS3022P100
Corning® reusable graduated cylinder
Z418838
Aldrich® flat-bottom flask
Z691461
Universal tube rack
USP
1370327
Lorazepam Related Compound A
A9539
琼脂糖
803410
Low Dead Volume Precolumn Filter with 0.5 μm Stainless Steel Frit
1.00467
GranuCult® plus 玫瑰-孟加拉氯霉素琼脂(RBC)
pkg of 1 ea
92 mm × 6.35 mm × 4.0 mm
正在寻找类似产品? 访问 产品对比指南
历史批次信息供参考:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our 文件 section.