21269-U
1 of 10
CLS4965B500
Pyrex® three-neck round-bottom flask
SYNF434400
Synthware™ three neck round bottom flask with angled side necks
Supelco
24056
SP®-2560毛细管GC色谱柱
Sigma-Aldrich
Z759317
Joint clip, standard taper
58298
SUPELCOSIL™ LC-18 HPLC 柱
28449-U
具有旋阀的顶部烧瓶
21266-U
收集烧瓶/烧杯
Millipore
XX2004720
Hydrosol 47 mm 过滤器支架,不锈钢
BR134080
BRAND®流量指示器
Z509450
Support stand with rod
pk of 3
solid phase extraction (SPE): suitable
250 mL
food and beverages
分析样品制备
样品处理
历史批次信息供参考:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our 文件 section.