主要文件
Z743322
Brayer for Breathe-EASY®) sealing membrane, 4 in firm deluxe rubber roller
pack of 1 ea
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41122410
NACRES:
NB.35
材料
rubber (soft)
包裝
pack of 1 ea
製造商/商標名
Diversified Biotech BRAY-5000
一般說明
Handy 4 inch soft rubber brayer ensures a good seal between microplate and sealing films.
法律資訊
Breathe-Easy is a registered trademark of Diversified Biotech
历史批次信息供参考:
分析证书(COA)