主要文件
Z740471
MICROGARD 2000 coveralls for protection from infective agent
size L (elasticated ankles)
登录查看公司和协议定价
About This Item
UNSPSC代码:
46181503
NACRES:
NB.35
尺寸
L (elasticated ankles)
制造商/商品名称
Arco 1762005
正在寻找类似产品? 访问 产品对比指南
一般描述
MICROGARD 2000 fully elasticated coverall with hood , setting the standard in Type 5/6 performance and protection, offers protection from infective agents, pesticides, particulates, fibres and liquid chemical spray.
Features:
  • 3-piece hood
  • 2-way front zipper with re-sealable storm flap
  • Finger loops
  • Elasticated hood, waist, wrists and ankles
Approved to:
EN ISO 13982-1 Type 5 Particulate protection
EN13034 Type 6 Reduced spray
EN1073-2 Barrier to radioactive particulates - Class 2
EN14126 Barrier to infective agents
EN1149-5 Anti-static
法律信息
Microgard is a trademark of Microgard Limited
历史批次信息供参考:
分析证书(COA)