Merck
Z640204
Eppendorf® epT.IPS 吸头盒
volume range 0.1-20 μL
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121600
NACRES:
NB.01
材料
medium gray rack
無菌
non-sterile
特點
barrier: no
包裝
pack of 96 (includes 1 reusable box)
製造商/商標名
Eppendorf® 0030 073.029
Eppendorf® 022491318
長度
40 mm
最大體積
20 μL
容積範圍
0.1-20 μL
正在寻找类似产品? 访问 产品对比指南
一般說明
ep T.I.P.S. 盒和钉仓系统的一部分,用于无污染地处理再灌注托盘,塑料盖保护托盘在转移到工作盒时不会接触托盘。
可高温高压消毒的工作箱,针对多道移液器进行了优化,带有可拆卸的盖子和更大的占地面积,避免翻倒。可堆叠。随附注意事项。
法律資訊
Eppendorf is a registered trademark of Eppendorf AG
访问文档库
商品
Optimized separation of Oligo Standard 6, an internally created HPLC-UV system suitability mix, on Chromolith® RP-18e columns with an evaluation of the effect of flow rates up to 3 mL/min and ion-pairing reagent, TEAA, on the separation.
A step-by-step protocol for sample preparation, calibration, digestion, and inductively coupled plasma-mass spectrometric (ICP-MS) analysis of heavy metals in six different hemp-containing beverages.
LC-UV-MS workflow details teriparatide peptide mapping, including enzymatic digestion, separation conditions, and QTOF mass spectrometer identification.
Analysis of heavy metals and metals in cannabis and hemp containing personal care products and cosmetics include sample preparation by grinding, preparation of calibration solutions for a standard addition approach, a microwave digestion protocol and subsequent ICP-MS analysis.
查看所有结果
实验方案
A complete SEC-UV workflow for quantifying monoclonal antibody fragments, monomers, and aggregates, including procedure for a system suitability test and heat stressing of antibodies.