主要文件
Z380024
Sigma-Aldrich
Thirsty Stix absorbants
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41100000
包裝
pkg of 250
存貨情形
not available in EU
長度 × 直徑
6 in. (150 mm) × 0.15 in. (3.8 mm)
應用
Adhesive-free, low-lint sticks of cylindrically wound filter paper are ideal for a multitude of uses. Remove liquid residues from sample tubes and supernatants from centrifuge tubes; Thirsty Stix absorb at least 100% of their weight in water, and 50-100% of other liquids. Use with solvent to clean laboratory apparatus and small orifices. Use as a sterile sponge, even as a paper chromatography column.
历史批次信息供参考:
分析证书(COA)