主要文件
Z374407
Sigma-Aldrich
Scienceware® microscope slide holder
accepts up to 10 single or double slides standing on end
登录查看公司和协议定价
About This Item
UNSPSC代码:
41122800
NACRES:
MA.01
特点
Holds 10 x slides 76x26 mm
包装
pack of 1 ea
制造商/商品名称
Bel-Art F44542-0000
直径 × 宽度 × 高度
10 in. (25 cm) × 2 in. (5 cm) × 1 1/4 in. (3 cm)
相关类别
一般描述
Slide holder has a sloped groove that accepts up to 10 single or double slides standing on end. Made of durable, extruded white plastic,
法律信息
Scienceware is a registered trademark of SP Scienceware, a division of SP
历史批次信息供参考:
分析证书(COA)