主要文件
Z360287
Sigma-Aldrich
Carboy spigots
polypropylene
别名:
plastic spigot, plastic spigots
登录查看公司和协议定价
About This Item
UNSPSC代码:
41121820
NACRES:
NB.35
物料
polypropylene
包装
pack of 1 ea
制造商/商品名称
Nalgene 6422-0010
正在寻找类似产品? 访问 产品对比指南
相关类别
一般描述
Spigots fit all Nalgene carboys. Both are autoclavable when installed on autoclavable carboys. Thread is 11/8 in. - 12 straight female threads. Spout accepts 5/8 in. I.D. tubing.
Adapter accepts 1/4 to 5/16 in. tubing. This spigot is the standard fitting for all Nalgene carboys. Do not autoclave adapter.
历史批次信息供参考:
分析证书(COA)