主要文件
Z329681
BRAND® wash bottle, wide-mouth, labelled, LDPE
capacity 250 mL, label, Methanol, without venting valve
登录查看公司和协议定价
About This Item
UNSPSC代码:
41103209
NACRES:
NB.17
物料
green closure
low-density polyethylene bottle
round bottle
translucent bottle
wide-mouth bottle
特点
label Methanol
with cap
without venting valve
包装
pack of 5 ea
制造商/商品名称
BRAND 144080
容量
250 mL
正在寻找类似产品? 访问 产品对比指南
一般描述
Flexible, high-quality wash bottles with integral wash steam and sealing cone in the closure. Large opening for convenient filling. Label with designation of contents and general safety statements (with CAS no. and NFPA codes).
Bottles with venting valve (metal ball) are for distilled water and organic solvents only; to avoid dropping caused by overpressure.
其他说明
BR144080 (India)
法律信息
BRAND is a registered trademark of BRAND GMBH + CO KG
配件
历史批次信息供参考:
分析证书(COA)