主要文件
Z324892
Sigma-Aldrich
BRAND® SILBERBRAND ETERNA volumetric pipettes, calibrated to deliver
capacity 100 mL
登录查看公司和协议定价
About This Item
UNSPSC代码:
41121500
NACRES:
NB.01
物料
AR-Glas®
Agency
according to DIN 12 690
特点
class B (callibrated to deliver)
class B
one-mark
包装
pack of 6 ea
制造商/商品名称
BRAND 29519
体积测量范围
100 mL, accuracy: 0.12 mL
容量
100 mL
正在寻找类似产品? 访问 产品对比指南
一般描述
The BRAND® SILBERBRAND ETERNA volumetric pipettes are expertly crafted fromAR-Glas® and conform to DIN EN ISO 648 standards, ensuring high precision and reliability. Calibrated to deliver (TD, Ex), these pipettes feature a waiting time of 5 seconds for optimal accuracy in liquid transfer. Each BLAUBRAND® bulb pipette is supplied with a batch number and includes one batch certificate per packing unit from the manufacturer, guaranteeing traceability and quality. This combination of quality materials and thorough documentation makes these pipettes an excellent choice for demanding laboratory environments.
特点和优势
  • BRAND® SILBERBRAND ETERNA, 1-mark, volumetric pipette is used for the accurate measurement of instruments for precision analyses.
  • Calibrated to deliver (TD, Ex).
其他说明
BR29519 (India)
法律信息
AR-Glas is a registered trademark of Schott AG
BRAND is a registered trademark of BRAND GMBH + CO KG
历史批次信息供参考:
分析证书(COA)
访问文档库
实验方案
本页面包含操作移液管和助吸器的详细信息。
This page contains detailed information on working with and handling pipettes and pipetting aids.