跳转至内容
Merck
所有图片(3)

主要文件

SAB5701530

Sigma-Aldrich

Anti-BNIP1 antibody produced in rabbit

登录查看公司和协议定价


About This Item

UNSPSC代码:
12352203
NACRES:
NA.41

生物来源

rabbit

质量水平

抗体产品类型

primary antibodies

克隆

monoclonal

表单

liquid

种属反应性

rat, human, mouse

浓度

0.3 mg/mL

技术

immunohistochemistry: 1:50-1:200
western blot: 1:500-1:2000

UniProt登记号

运输

wet ice

储存温度

−20°C

靶向翻译后修饰

unmodified

基因信息

human ... BNIP1(662)

一般描述

This gene is a member of the BCL2/adenovirus E1B 19 kd-interacting protein (BNIP) family. It interacts with the E1B 19 kDa protein, which protects cells from virally-induced cell death. The encoded protein also interacts with E1B 19 kDa-like sequences of BCL2, another apoptotic protector. In addition, this protein is involved in vesicle transport into the endoplasmic reticulum. Alternative splicing of this gene results in four protein products with identical N- and C-termini. [provided by RefSeq, Mar 2011]

免疫原

A synthesized peptide derived from human BNIP1

外形

Buffer: PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

未找到合适的产品?  

试试我们的产品选型工具.

储存分类代码

12 - Non Combustible Liquids

WGK

WGK 1

闪点(°F)

Not applicable

闪点(°C)

Not applicable


历史批次信息供参考:

分析证书(COA)

Lot/Batch Number

没有发现合适的版本?

如果您需要特殊版本,可通过批号或批次号查找具体证书。

已有该产品?

在文件库中查找您最近购买产品的文档。

访问文档库

我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.

联系技术服务部门