Merck
RF454811P-EP
Rotor for Eppendorf® 5804/5804R & 5810/5810R Centrifuges
FA-45-48-PCR, to hold, 6 x 8-tube or 5-tube PCR strips, to hold, 48 x 0.2 mL PCR tubes
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41103909
特點
to hold 48 x 0.2 mL PCR tubes
to hold 6 x 8-tube or 5-tube PCR strips
fixed angle strip rotor
包裝
pack of 1 ea
製造商/商標名
Eppendorf® 022638581 (NA)
Eppendorf® 5804735005 (EU)
參數
15,350 × g max. RCF
技術
PCR: suitable
centrifugation: suitable
相容性
for use with centrifuge models 5804/5804R/5810/5810R
正在寻找类似产品? 访问 产品对比指南
法律資訊
Eppendorf is a registered trademark of Eppendorf AG