主要文件
OH83033969
Ohaus® pH Electrode
STPURE
登录查看公司和协议定价
About This Item
UNSPSC代码:
41115604
包装
pack of 1 ea
制造商/商品名称
Ohaus® 83033969
pH值测量范围
0.00-14 pH unit
正在寻找类似产品? 访问 产品对比指南
一般描述
All electrodes include a temperature sensor, which powers automatic temperature compensation and ensures accurate measurements.
应用
pH, reference, oxidation-reduction potential (ORP), conductivity, dissolved oxygen (DO), temperature measurement
特点和优势
Can be used in conjunction with all Starter bench and portable meters
法律信息
Ohaus is a registered trademark of Ohaus Corp.
历史批次信息供参考:
分析证书(COA)