主要文件
OH83033962
Ohaus® pH Meter
Model ST300-B
登录查看公司和协议定价
About This Item
UNSPSC代码:
41115603
物料
epoxy shaft
包装
pack of 1 ea
制造商/商品名称
Ohaus® 83033962
ORP measuring range
-1999-+1999 mV
pH值测量范围
0.00-14.00 pH unit
温度测量范围
0-100 °C
电池电量
4 AAA, 250 hr life
正在寻找类似产品? 访问 产品对比指南
一般描述
Easy-to-use and accurate with a simple calibration process, automatic temperature compensation and fast results. Ideal for secure field use with a meter stand, wrist strap and durable IP54 housing.
应用
pH, oxidation-reduction potential (ORP) measurement
特点和优势
Automatic and manual temperature compensation, auto buffer recognition, calibration icon
法律信息
Ohaus is a registered trademark of Ohaus Corp.
历史批次信息供参考:
分析证书(COA)