生物源
goat
共軛
unconjugated
抗體表格
affinity isolated antibody
抗體產品種類
primary antibodies
無性繁殖
polyclonal
形狀
lyophilized powder
物種活性
mouse
技術
indirect ELISA: 0.5-1.0 μg/mL
neutralization: suitable
western blot: 0.1-0.2 μg/mL
UniProt登錄號
儲存溫度
−20°C
基因資訊
mouse ... Csf1(12977)
一般說明
Macrophage Colony Stimulating Factor (M-CSF) is a cytokine that plays a crucial role in host response to viral and fungal infections. It is secreted by fibroblasts, monocytes/macrophages and epithelial cells. M-CSF has been reported to the growth and differentiation of monocytes/macrophages. The M-CSF levels increase rapidly in response to fungal infections by Candida albicans and Aspergillus fumigatus. Reports suggest that M-CSF may act in an autocrine/paracrine manner to sustain HIV replication and increase the frequency by which the macrophages become infected with the virus. M-CSF may be a treatment option to combat infections associated with neutropenia. Anti-Macrophage Colony Stimulating Factor antibody specifically reacts with mouse M-CSF. The antibody shows approximately 5% cross-reactivity with recombinant human M-CSF.
特異性
By immunoblotting and ELISA, the antibody shows 5% cross-reactivity with recombinant human M-CSF.
免疫原
recombinant mouse M-CSF expressed in Escherichia coli.
應用
Anti-Macrophage Colony Stimulating Factor antibody may be used for ELISA at a working antibody concentration of 0.5-1.0 μg/ml. For immunoblotting the recommended concentration is 0.1-0.2 μg/ml. It is also suitable to neutralize the biological activity of mouse M-CSF in neutralization assays at a working concentration of 0.015-0.035 μg/ml. The antibody was used to neutralize M-CSF of murine embryonic fibroblasts.
外觀
Lyophilized from a 0.2 μm-filtered solution of phosphate-buffered saline.
未找到合适的产品?
试试我们的产品选型工具.
Cell reports. Medicine, 2(12), 100457-100457 (2022-01-15)
Second generation (2G) chimeric antigen receptors (CARs) contain a CD28 or 41BB co-stimulatory endodomain and elicit remarkable efficacy in hematological malignancies. Third generation (3G) CARs extend this linear blueprint by fusing both co-stimulatory units in series. However, clinical impact has
Journal of leukocyte biology, 82(4), 915-925 (2007-07-27)
G-CSF and GM-CSF play important roles in regulating neutrophil production, survival, differentiation, and function. However, we have shown previously that G-CSF/GM-CSF double-deficient [knockout (KO)] mice still develop a profound neutrophilia in bone marrow and blood after infection with Candida albicans.
Journal of immunology (Baltimore, Md. : 1950), 154(10), 5528-5535 (1995-05-15)
Human monocyte-derived macrophages (MDM) cultured in medium containing macrophage (M) CSF are more susceptible to infection with HIV-1. M-CSF increases the frequency with which MDM become infected, the level of HIV mRNA expressed per infected cell, and the level of
The Journal of infectious diseases, 172(4), 1028-1034 (1995-10-01)
The effects of recombinant human macrophage colony-stimulating factor (M-CSF) on antifungal activity of human monocytes (MNC), MNC-derived macrophages (MDM), and rabbit pulmonary alveolar macrophages (PAM) against Aspergillus fumigatus were studied. MNC-induced hyphal damage was augmented by incubation with M-CSF (P
Infection and immunity, 59(3), 868-872 (1991-03-01)
Serum and tissue concentrations of the macrophage-specific colony-stimulating factor (CSF-1) and the number of CSF-1-responsive cells in bone marrow were investigated in mice chronically infected with a low-virulence strain of the opportunistic zoopathogenic yeast Candida albicans. CSF-1 levels in serum
我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.
联系技术服务部门