主要文件
G6266
Sigma-Aldrich
Guards for Sigma-Aldrich® pH electrodes
For 120 mm stem length
登录查看公司和协议定价
About This Item
UNSPSC代码:
41115603
NACRES:
NB.79
一般描述
Removable plastic guard that protects the glass bulb from impact during use.
法律信息
SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany
历史批次信息供参考:
分析证书(COA)