主要文件
EPS44I041006
Eppendorf® Mandatory stacking kit; Innova S44i, to allow installation
登录查看公司和协议定价
About This Item
UNSPSC代码:
41103800
包装
pkg of 1 ea
制造商/商品名称
Eppendorf® S44I041006
正在寻找类似产品? 访问 产品对比指南
一般描述
The Eppendorf Stacking Kit is designed for the triple stacking of the Innova S44i shakers, enabling users to maximize lab space and increase productivity. This stacking kit provides a secure and stable platform for placing a second shaker on top of the first S44i unit. By utilizing this kit, researchers can optimize their work area without sacrificing performance. Easy to install and compatible with the S44i model, this stacking kit is an ideal solution for laboratories aiming to streamline their operations efficiently.
其他说明
Maximum agitation speed of Innova S44i reduced to 300 rpm when stacked on one Innova 44/44R and 250 rpm when triple stacked
法律信息
Eppendorf is a registered trademark of Eppendorf AG
历史批次信息供参考:
分析证书(COA)