Merck
EP5920769006
Eppendorf® Adapter for 7 x 10 ml Oak Ridge tubes
for FA-6x250 rotor, case of 2 ea
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41103911
NACRES:
NB.82
特點
for FA-6x250 rotor
包裝
case of 2 ea
製造商/商標名
Eppendorf® 5920769006
參數
10,000 rpm speed
儲存溫度
room temp
法律資訊
Eppendorf is a registered trademark of Eppendorf AG