Key Documents
EP5490039009
Rotor for Eppendorf® Concentrator Plus
F-35-8-24, non aerosol tight, to hold, 8 x 25 mL flat-bttom tubes
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41103909
NACRES:
NB.82
特點
to hold 8 x 24 x 86 -90 mm
to hold 8 x 25 mL flat-bttom tubes
fixed angle rotor
non aerosol tight
包裝
pack of 1 ea
製造商/商標名
Eppendorf® 5490039009 (EU)
技術
centrifugation: suitable
相容性
for use with centrifuge models Concentrator Plus
正在寻找类似产品? 访问 产品对比指南
法律資訊
Eppendorf is a registered trademark of Eppendorf AG