Merck
EP5427732004
Eppendorf® Adapter
holds 1 x 7-15 mL, bore size (small), pack of 2 ea
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41103911
NACRES:
NB.82
特點
holds 1 x 7-15 mL
包裝
pack of 2 ea
製造商/商標名
Eppendorf® 5427732004
技術
centrifugation: suitable
孔尺寸
(small)
相容性
for use with Rotor F-35-6-30 (small rotor bore)
法律資訊
Eppendorf is a registered trademark of Eppendorf AG