主要文件
EP5392070038
Eppendorf(R) Plate Adapter HeatSealer S200
for 96 and 384 well plates
登录查看公司和协议定价
About This Item
UNSPSC代码:
41121815
NACRES:
NB.15
特点
for 96 and 384 well plates
制造商/商品名称
Eppendorf® 5392070038 (Global)
正在寻找类似产品? 访问 产品对比指南
相关类别
特点和优势
Hermetic Sealing, of multiwell plates
Eliminates evaporation in PCR
Seals 96- and 384-well plates
Integrated thermostat prevents overheating
Improved mechanics for even easier Sealing,
PCR Plate Adapter included
法律信息
Eppendorf is a registered trademark of Eppendorf AG
历史批次信息供参考:
分析证书(COA)