EP5353040121
1 of 10
EP0030119800
Eppendorf® Tube Rack for 0.5 mL Tubes
CLS3749
Corning® Thermowell GOLD PCR tubes
HS29025G
Microtube Rack
EP0030119819
Eppendorf® Tube Rack for 1.5 and 2.0 mL Tubes
Z756482
Microcentrifuge tube-PCR rack, 80 well, reversible
Sigma-Aldrich
R5651
Four-way microtube rack
HS4323
Microcentrifuge Tube
Z743197
Julabo® bath test tube racks
CLS4365
Corning® 聚苯乙烯泡沫塑料管架(50 mL 离心管)
Z765503
Benchmixer™ XL multi-tube vortexer
Holds 24 x 0.5mL micro test tubes
Eppendorf® 5353040121