EP4920620006
1 of 10
EP0030140524
Storage Box for Eppendorf® Tubes
EP0030089685
Eppendorf® Combitips Advanced®
EP0030089677
EP0030122364
Eppendorf® Tubes 5 mL
EP4987000134
Eppendorf® Repeater™ E3/E3x
EP0030089456
EP0030122321
Eppendorf® Tubes 5.0 mL
EP0030122305
EP4861000155
Eppendorf® Xplorer® 12-channel pipettor
Z177814
Scienceware® polypropylene trays
color code: violet variable
pkg of 1 ea
Eppendorf® 4920620006
5 mL
正在寻找类似产品? 访问 产品对比指南
历史批次信息供参考:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our 文件 section.