Merck
CLS431465
Corning® flask, canted neck, 150 cm, vented
别名:
cell culture flasks, tissue culture flasks
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41122104
NACRES:
NB.13
材料
canted neck
high-density polyethylene cap
polystyrene
rectangular bottom flask
vent cap
無菌
sterile; γ-irradiated
特點
cap
closure type Vented cap
包裝
pack of 5
case of 50
製造商/商標名
Corning 431465
表面積
150 cm2
有效容積
30-45 mL
結合類型
non-treated surface
相关类别
法律資訊
Corning is a registered trademark of Corning, Inc.