主要文件
CLS2679840
Corning® polypropylene closure
fits neck O.D. 40 mm
, natural
登录查看公司和协议定价
About This Item
UNSPSC代码:
41121800
物料
natural
polypropylene
包装
case of 100
正在寻找类似产品? 访问 产品对比指南
相关类别
一般描述
For culture tubes and flasks with long rimless necks. Position closure open for gas exhange or closed for humidified environment. Internal drip ring minimizes contamination.
法律信息
Corning is a registered trademark of Corning, Inc.
历史批次信息供参考:
分析证书(COA)