AXYP96450VBK
别名:
96 multiwell plate
V-bottom black wellsblack plate
non-sterile
Axygen Inc P-96-450V-BKCorning
pkg of 10x5plates/cs
500 μL
96 wells
450 μL
suitable for PCR
non-treated surface
正在寻找类似产品? 访问 产品对比指南
Corning®
实验室容器与储存
实验室耗材合作伙伴
历史批次信息供参考:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our 文件 section.