主要文件
ADAPA438107-EP
Adapter for Eppendorf® A-4-38 Rotor
holds 10 x 5-7 mL round-bottom tubes, for use with 90 mL rectangular bucket, pkg of 2 ea
登录查看公司和协议定价
About This Item
UNSPSC代码:
41103911
特点
holds 10 x 5-7 mL round-bottom tubes
包装
pack of 2 ea
pkg of 2 ea
制造商/商品名称
Eppendorf® 022639323
技术
centrifugation: suitable
相容性
for use with 90 mL rectangular bucket
一般描述
Adapter to be used with 90 mL rectangular bucket in Eppendorf A-4-38 rotor. Holds 10 x 5-7 mL round-bottom tubes and supplied as a pack of 2.
法律信息
Eppendorf is a registered trademark of Eppendorf AG
历史批次信息供参考:
分析证书(COA)