主要文件
1336R
Millipore
BIOEAZE Rigging Sets
登录查看公司和协议定价
About This Item
UNSPSC代码:
12352200
描述
Designed for use with Quick Connect Applications with Male and Female Quick Connects with Caps, Double Polybagged
无菌性
sterile; γ-irradiated
正在寻找类似产品? 访问 产品对比指南
一般描述
1 x 10 ft. length of Size 15 Opaque 001 C-Flex® Tubing and 1/4" Quick Connects with Caps
包装
双层塑料袋装
法律信息
BIOEAZE is a trademark of Sigma-Aldrich Co. LLC
历史批次信息供参考:
分析证书(COA)