推荐产品
运输
dry ice
储存温度
−20°C
SMILES字符串
[N-]=[N+]=Cc1c2ccccc2cc3ccccc13
应用
Common fluorescent diazomethane analogue used to derivatize biomolecules. Also has been used to detect several types of acids, including amino acids, bile acids, fatty acids, prostaglandins, and steroid acids.
警示用语:
Warning
危险声明
危险分类
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
靶器官
Respiratory system
储存分类代码
11 - Combustible Solids
WGK
WGK 3
闪点(°F)
Not applicable
闪点(°C)
Not applicable
个人防护装备
dust mask type N95 (US), Eyeshields, Gloves
Sheng-Feng Lai et al.
Journal of nanobiotechnology, 13, 85-85 (2015-11-22)
Nanoparticles can be used for targeted drug delivery, in particular for brain cancer therapy. However, this requires a detailed analysis of nanoparticles from the associated microvasculature to the tumor, not easy because of the required high spatial resolution. The objective
T H Roemen et al.
Journal of chromatography, 570(2), 243-251 (1991-10-04)
A high-performance liquid chromatographic technique for the rapid assessment fatty acids in cardiac tissue is described. A level of 50.4 +/- 14.9 nmol fatty acids per g wet weight of rat myocardial tissue could be monitored. The content of the
R Draisci et al.
Journal of chromatography. A, 798(1-2), 137-145 (1998-05-02)
A new analogue of okadaic acid (OA), the toxin mainly responsible for diarrhetic shellfish-poisoning (DSP) phenomena in Europe, has been isolated from toxic phytoplankton (Dinophysis acuta) collected in Irish waters. Fluorimetric LC analyses of the extracts of bulk phytoplankton samples
P Demin et al.
Journal of chromatography. B, Biomedical applications, 672(2), 282-289 (1995-10-20)
Fluorescent anthryl (ADAM) derivatives of hepoxilins have been shown to possess good chromatographic properties affording good sensitivity for the high-performance liquid chromatographic analysis and detection of these compounds and related eicosanoids (12-hydroxyeicosatetraenoic acid) in biological samples. We report herein the
O B Stabell et al.
Toxicon : official journal of the International Society on Toxinology, 29(1), 21-29 (1991-01-01)
Okadaic acid and dinophysistoxin-1, the principal toxic components in diarrhetic shellfish poisoning, may be detected by high-performance liquid chromatography and fluorometric measurement as 9-anthrylmethyl esters. However, "greasy" samples may occur and the fluorescent reagent 9-anthryldiazomethane may decompose during storage, resulting
我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.
联系技术服务部门