跳转至内容
Merck

51686

Sigma-Aldrich

4-Heptadecylumbelliferone

for fluorescence, ≥98.0% (TLC)

别名:

4-Heptadecyl-7-hydroxycoumarin

登录查看公司和协议定价


About This Item

经验公式(希尔记法):
C26H40O3
CAS号:
分子量:
400.59
Beilstein:
1263899
MDL號碼:
分類程式碼代碼:
12352200

等級

for fluorescence

化驗

≥98.0% (TLC)

mp

95 °C (lit.)

溶解度

DMF: soluble
DMSO: soluble

螢光

λex 367 nm; λem 450 nm in 0.1 M Tris pH 9.0

SMILES 字串

CCCCCCCCCCCCCCCCCC1=CC(=O)Oc2cc(O)ccc12

應用

4-Heptadecylumbelliferone (HUF, C17-HC), a pH-sensitive fluorophore, is used to study fluorescence anisotropy, electrical surface potential and pH of cell membranes and lipoid structures such as emulsions and micelles.

包裝

Bottomless glass bottle. Contents are inside inserted fused cone.

其他說明

Lipoid pH indicator

象形圖

Exclamation mark

訊號詞

Warning

危險聲明

危險分類

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

儲存類別代碼

11 - Combustible Solids

水污染物質分類(WGK)

WGK 3

閃點(°F)

Not applicable

閃點(°C)

Not applicable

個人防護裝備

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

分析证书(COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

已有该产品?

在文件库中查找您最近购买产品的文档。

访问文档库

Sony George et al.
The journal of physical chemistry. B, 113(15), 5117-5127 (2009-03-26)
Steady-state and time-resolved fluorescence anisotropy measurements using probes coumarin 153 (C153) and 4-heptadecylumbelliferon (HUF) have been carried out to understand the micelle to gel transition of an aqueous triblock copolymer P123 ((EO)(20)-(PO)(70)-(EO)(20)) (EO = ethylene oxide; PO = propylene oxide)
Laura Rabinovich-Guilatt et al.
Chemistry and physics of lipids, 131(1), 1-13 (2004-06-24)
The present study is aimed to characterize the electrostatic parameters of oil in water emulsion droplets composed of MCT (medium chain triglycerides), PL (phospholipids) and Poloxamer and containing increasing concentrations of the cationic lipid oleylamine (OA), in Hepes 20 mM
P J Dykes et al.
The British journal of dermatology, 110(3), 283-292 (1984-03-01)
Mechanical stimulation of the skin surface in the presence of non-ionic detergent (Triton X-100)-containing buffers has been used to prepare isolated corneocytes and non-ionic detergent-soluble (NIDS) material from human stratum corneum. Analysis of the major lipid classes present in the
C.J. Drummond et al.
Photochemistry and Photobiology, 45, 19-19 (1987)
C Saldanha et al.
The Journal of membrane biology, 190(1), 75-82 (2002-11-08)
An experimental approach has been developed to study human erythrocyte vesiculation, using the fluorescent probes diphenylhexatriene (DPH), trimethylamino-diphenylhexatriene (TMA-DPH) and heptadecyl-hydroxycoumarin (C17-HC). Acetylcholinesterase (AChE) enzyme activity measurements confirmed the presence of exovesicles released from erythrocyte membranes labeled with DPH, TMA-DPH

我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.

联系技术服务部门