主要文件
ZSTWIN0A1
Milli-Q
Tank Level Adapter
Transmits tank pure water feeding level information to Milli-Q® IQ 7000 and EQ 7000 systems
登录查看公司和协议定价
About This Item
UNSPSC代码:
41104214
NACRES:
JA.13
包装
pkg of 1 unit
相容性
for use with Milli-Q® EQ 7000
for use with Milli-Q® IQ 7000
一般描述
This electronic sensor transfers tank level information to the Milli-Q® IQ 7000 and Milli-Q® EQ 7000 water purification systems. It stops water feed from an empty tank and lets you check water levels in the tank before sourcing water, preventing air from entering the system.
应用
Transmits the polyethylene (PE) tank pure water feeding level information to Milli-Q® IQ 7000 and Milli-Q® EQ 7000 ultrapure water purification systems.
法律信息
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany