P83075
1 of 10
Millipore
P83071
Integritest 4打印纸
IT5INS001
Integritest® 5过滤器完整性测试仪
XIT4TPRTP3
Integritest热敏纸
Sigma-Aldrich
163M-1
CD163 (MRQ-26) Mouse Monoclonal Antibody
IT5SP0014
F1 过滤器组件,水阱
XX42P0080
Pellicon®不锈钢支架
MP0DDSPMAN1
Millistak+® Pod Disposable Manifold
P83076
进气管空气过滤器,带干燥剂(Inlet Tubing w/Dessicant Air Filter)
XX42T1901
Tri-Clover Gaskets 3/4 in. Silicone
103R-9
CD3 (MRQ-39) Rabbit Monoclonal Antibody
Integritest®
for use with INTEGRITEST® 4
Integrity Testers