特點
mercury-free UV LED
包裝
pkg of 1 unit
製造商/商標名
ech2o®
參數
265 nm (wavelength)
相容性
for use with Milli-Q® IQ 7003
for use with Milli-Q® IQ 7005
for use with Milli-Q® IQ 7010
for use with Milli-Q® IQ 7015
一般說明
ECH2O® ASM灯通过定期暴露在杀菌紫外线下保护储水的完整性。该ECH2O® ASM灯是无汞UV LED (265 nm),可防止Milli-Q® IQ/IX储罐内表面的细菌生长和生物膜形成。
應用
它可以防止储罐内部的生物膜形成和细菌增殖。
法律資訊
ECH2O is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany