推荐产品
特點
CE compliant
包裝
pkg of 1 ea
製造商/商標名
Bollé COVACLAVE
存貨情形
not available in USA
一般說明
Specific ultra-comfort goggle for clean rooms. The Bollé Coverall goggle will assure you the best protection and the best comfort thanks to its interchangeable ventilation and with the possibility to wear prescription spectacles underneath.
- Ultra-comfort autoclavable goggle
- Fits over prescription spectacles
- Upper indirect and interchangeable ventilation
- Adjustable strap
- Uncoated polycarbonate lens