主要文件
Z758957
Bollé coverall autoclave safety goggles for clean rooms
CE compliant
别名:
protective eyewear, protective glasses, safety eyeglasses, safety eyewear, safety glasses
登录查看公司和协议定价
About This Item
分類程式碼代碼:
46181802
特點
CE compliant
包裝
pkg of 1 ea
製造商/商標名
Bollé COVACLAVE
存貨情形
not available in USA
一般說明
Specific ultra-comfort goggle for clean rooms. The Bollé Coverall goggle will assure you the best protection and the best comfort thanks to its interchangeable ventilation and with the possibility to wear prescription spectacles underneath.
  • Ultra-comfort autoclavable goggle
  • Fits over prescription spectacles
  • Upper indirect and interchangeable ventilation
  • Adjustable strap
  • Uncoated polycarbonate lens
从最新的版本中选择一种:
分析证书(COA)