主要文件
Z742520
Block for digital dry bath and MyBlock
Holds 12 x 15 mL tubes
登录查看公司和协议定价
About This Item
UNSPSC代码:
41102404
NACRES:
NB.77
特点
Holds 12 x 15 mL tubes
包装
pack of 1 ea
制造商/商品名称
Benchmark Scientific BSW15
正在寻找类似产品? 访问 产品对比指南
相关类别
一般描述
For use with Benchmark Digital Dry Bath
  • Material: high grade aluminum
  • Precision machined to match the tube shape and size, providing unsurpassed temperature uniformity
法律信息
MyBlock is a trademark of Benchmark Scientific, Inc.
历史批次信息供参考:
分析证书(COA)