主要文件
Z740135
QianCap pressure monitor-25
for tubes/bottles with internal threads #25
登录查看公司和协议定价
About This Item
UNSPSC代码:
41100000
NACRES:
NB.51
包装
pkg of 1 ea
制造商/商品名称
Q Labtech
参数
0-300 psi pressure range
一般描述
QianCap pressure monitor-25 is designed for monitoring internal pressure on pressure tubes, flasks and bottles with #25 internal threads. The set contains all items needed for immediate use (pressure release system, 1 PTFE adapter-25H, 1 Simriz O-ring-25-S, 1 stainless steel pressure gauge-300 psi and 10 septa-12 mL).
特点和优势
  • Monitors Ace-Thred pressure tubes/bottles.
  • Provides optional higher pressure adapter (120 psi).
法律信息
QianCap is a trademark of Q Labtech LLC
历史批次信息供参考:
分析证书(COA)