主要文件
Z682896
Ace receiving flask,
capacity 3000 mL, joint: ST/NS 35/20, plastic-coated glass
登录查看公司和协议定价
About This Item
UNSPSC代码:
41121804
NACRES:
NB.43
物料
plastic-coated glass
round bottom flask
single-neck
接合处
joint: ST/NS35/20
无菌性
non-sterile
特点
closure type Standard Taper Joint
包装
case of 1
制造商/商品名称
Büchi equivalent, 40777/25266
容量
3000 mL
正在寻找类似产品? 访问 产品对比指南
相关类别
一般描述
The receiving flasks have the standard joint to fit the glassware used in rotary evaporators, and are available with poly-coating or plain glass.
历史批次信息供参考:
分析证书(COA)